missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Butylphenylazo)phenol, TCI America™

$152.78 - $432.38

Chemical Identifiers

CAS 2496-21-1
Molecular Formula C16H18N2O
Molecular Weight (g/mol) 254.333
MDL Number MFCD00430795
InChI Key MHSQAHVLQVTXKH-UHFFFAOYSA-N
Synonym 4-Butyl-4′-hydroxyazobenzene
PubChem CID 5705751
IUPAC Name 4-[(4-butylphenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
SMILES CCCCC1=CC=C(C=C1)NN=C2C=CC(=O)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23575G View Documents TCI America
B23575G
5g Each for $152.78
 
B235725G View Documents TCI America
B235725G
25g Each for $432.38
 
Specifications

Chemical Identifiers

2496-21-1
254.333
MHSQAHVLQVTXKH-UHFFFAOYSA-N
5705751
CCCCC1=CC=C(C=C1)NN=C2C=CC(=O)C=C2
C16H18N2O
MFCD00430795
4-Butyl-4′-hydroxyazobenzene
4-[(4-butylphenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one

Specifications

Undesignated
5g
C16H18N2O
4-Butyl-4′C-hydroxyazobenzene
CCCCC1=CC=C(C=C1)NN=C2C=CC(=O)C=C2
254.333
254.33
≥98.0% (GC)
80°C
2496-21-1
MFCD00430795
MHSQAHVLQVTXKH-UHFFFAOYSA-N
4-[(4-butylphenyl)hydrazinylidene]cyclohexa-2,5-dien-1-one
5705751
Crystal-Powder at 20°C
4-(4-Butylphenylazo)phenol
Safety and Handling

Safety and Handling

TSCA : No