missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[4-(Bromomethyl)phenyl]-2-methyl-1,3-thiazole, 95%, Maybridge

Chemical Identifiers

CAS 857283-95-5
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C11H10BrNS
Formula Weight 268.18
MDL Number MFCD07772835
Chemical Name or Material 4-[4-(Bromomethyl)phenyl]-2-methyl-1, 3-thiazole
Assay 95%
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC40308CB SDS Fisher Scientific Company
CC40308CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

857283-95-5
100.0
C11H10BrNS
MFCD07772835
95%
94.5
95%
268.18
4-[4-(Bromomethyl)phenyl]-2-methyl-1, 3-thiazole
Technical Grade

Specifications

857283-95-5
100.0
250mg
C11H10BrNS
MFCD07772835
95%
94.5
Amber Glass Bottle
95%
268.18
4-[4-(Bromomethyl)phenyl]-2-methyl-1, 3-thiazole
Technical Grade

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger