missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[4-(Bromomethyl)phenyl]-1,2,3-thiadiazole, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC16408CB

 View more versions of this product

Catalog No. CC16408CB



Description

Description

Specifications

Specifications

4-[4-(Bromomethyl)phenyl]-1, 2, 3-thiadiazole
250mg
96.5
97%
MFCD00052102
DGHQOPZIGDRUIT-UHFFFAOYSA-N
4-[4-(bromomethyl)phenyl]thiadiazole
2776454
97%
Amber Glass Bottle
163798-92-3
100.0
C9H7BrN2S
1,2,3-thiadiazole,4-4-bromomethyl phenyl, 4-1,2,3-thiadiazol-4-yl benzyl bromide, 4-4-bromomethyl phenyl thiadiazole, 4-4-bromomethyl phenyl-1,2,3-thiadiazole, 4-4-bromomethyl-phenyl-1,2,3 thiadiazole, 4-4-bromomethylphenyl-1,2,3-thiadiazole
BrCC1=CC=C(C=C1)C1=CSN=N1
255.13
255.14

Chemical Identifiers

163798-92-3
255.13
DGHQOPZIGDRUIT-UHFFFAOYSA-N
2776454
BrCC1=CC=C(C=C1)C1=CSN=N1
C9H7BrN2S
MFCD00052102
1,2,3-thiadiazole,4-4-bromomethyl phenyl, 4-1,2,3-thiadiazol-4-yl benzyl bromide, 4-4-bromomethyl phenyl thiadiazole, 4-4-bromomethyl phenyl-1,2,3-thiadiazole, 4-4-bromomethyl-phenyl-1,2,3 thiadiazole, 4-4-bromomethylphenyl-1,2,3-thiadiazole
4-[4-(bromomethyl)phenyl]thiadiazole
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents
Product Certifications