Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Bromo-2-fluorobenzyl)morpholine, 96%

Chemical Identifiers

Chemical Name or Material 4-(4-Bromo-2-fluorobenzyl)morpholine
CAS 338454-98-1
Formula Weight 274.13
MDL Number MFCD12022322
Synonym 4-[(4-bromo-2-fluorophenyl)methyl]morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6271603

 
alfa aesar™
H6271603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6271606

 
alfa aesar™
H6271606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62716MD

 
alfa aesar™
H62716MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.13
Chemical Name or Material 4-(4-Bromo-2-fluorobenzyl)morpholine
Physical Form Solid
Assay 96%
Synonym 4-[(4-bromo-2-fluorophenyl)methyl]morpholine
CAS 338454-98-1
Quantity 1g
MDL Number MFCD12022322