missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-bromo-1,3-thiazol-2-yl)morpholine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1017781-60-0
Molecular Formula C7H9BrN2OS
Molecular Weight (g/mol) 249.126
MDL Number MFCD09607696
InChI Key XPFAGXZWVZAYCS-UHFFFAOYSA-N
Synonym 4-4-bromo-1,3-thiazol-2-yl morpholine, 4-4-bromo-thiazol-2-yl-morpholine, 4-4-bromothiazol-2-yl morpholine, 4-bromo-2-morpholin-4-yl-1,3-thiazole, 4-bromo-2-morpholinothiazole, acmc-2097ww, morpholine, 4-4-bromo-2-thiazolyl, morpholine,4-4-bromo-2-thiazolyl
PubChem CID 43811042
IUPAC Name 4-(4-bromo-1,3-thiazol-2-yl)morpholine
SMILES C1COCCN1C2=NC(=CS2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC62810DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC62810DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC62810DE
SDS View Documents Promotion Details
Fisher Scientific Company
CC62810DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1017781-60-0
249.126
XPFAGXZWVZAYCS-UHFFFAOYSA-N
43811042
C1COCCN1C2=NC(=CS2)Br
C7H9BrN2OS
MFCD09607696
4-4-bromo-1,3-thiazol-2-yl morpholine, 4-4-bromo-thiazol-2-yl-morpholine, 4-4-bromothiazol-2-yl morpholine, 4-bromo-2-morpholin-4-yl-1,3-thiazole, 4-bromo-2-morpholinothiazole, acmc-2097ww, morpholine, 4-4-bromo-2-thiazolyl, morpholine,4-4-bromo-2-thiazolyl
4-(4-bromo-1,3-thiazol-2-yl)morpholine

Specifications

1017781-60-0
100.0
C7H9BrN2OS
Amber Glass Bottle
1g
C1COCCN1C2=NC(=CS2)Br
249.126
249.13
4-(4-Bromo-1, 3-thiazol-2-yl)morpholine
96.5
97%
MFCD09607696
4-4-bromo-1,3-thiazol-2-yl morpholine, 4-4-bromo-thiazol-2-yl-morpholine, 4-4-bromothiazol-2-yl morpholine, 4-bromo-2-morpholin-4-yl-1,3-thiazole, 4-bromo-2-morpholinothiazole, acmc-2097ww, morpholine, 4-4-bromo-2-thiazolyl, morpholine,4-4-bromo-2-thiazolyl
XPFAGXZWVZAYCS-UHFFFAOYSA-N
4-(4-bromo-1,3-thiazol-2-yl)morpholine
43811042
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications