Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 97%

$127.76 - $375.58

Chemical Identifiers

Chemical Name or Material 4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
CAS 864754-13-2
Molecular Formula C22H33BN2O5
Formula Weight 416.32
MDL Number MFCD08706021
Synonym tert-Butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2890506

 
alfa aesar™
H2890506
5g Each for $375.58

AAH2890503

 
alfa aesar™
H2890503
1g Each for $127.76
Description & Specifications

Specifications

Formula Weight 416.32
Chemical Name or Material 4-(4-Boc-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C22H33BN2O5
MDL Number MFCD08706021
CAS 864754-13-2
Synonym tert-Butyl 4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]piperazine-1-carboxylate
Quantity 5g