Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Bis(diphenylphosphinoyl)-2,2',6,6'-tetramethoxy-3,3'-bipyridine, 95%

$460.68 - $460.68

Chemical Identifiers

Chemical Name or Material 4,4'-Bis(diphenylphosphinoyl)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
CAS 220998-37-8
Molecular Formula C38H34N2O6P2
Formula Weight 676.63g/mol
MDL Number MFCD12546201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3144106

 
alfa aesar™
H31441-06
5g Each for $460.68
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material 4,4'-Bis(diphenylphosphinoyl)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
CAS 220998-37-8
Assay 95%
Molecular Formula C38H34N2O6P2
Formula Weight 676.63g/mol
MDL Number MFCD12546201
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.