Please login to your online account to display your discounted pricing

4,4'-Bis(dimethylamino)benzil, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  402390050

 View more versions of this product

Catalog No. AC402390050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (HPLC)
Physical Form Crystalline Powder
Formula Weight 296.36
Quantity 5g
Color Yellow-green to green
Melting Point 200° to 204°C
Chemical Name or Material 4, 4'-Bis(dimethylamino)benzil
Infrared Spectrum Authentic
Molecular Formula C18H20N2O2
Packaging Glass bottle
Assay Percent Range 98%
CAS 17078-27-2

Specifications

Chemical Name or Material 4, 4'-Bis(dimethylamino)benzil
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C18H20N2O2
Formula Weight 296.36