missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Bis(dimethylamino)benzil, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC402390250

 View more versions of this product

Catalog No. AC402390250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4, 4'-Bis(dimethylamino)benzil
Authentic
Crystalline Powder
98%
296.36
200° to 204°C
17078-27-2
Glass bottle
≥97.5% (HPLC)
C18H20N2O2
Yellow-green to green
25g

Chemical Identifiers

98%
296.36
≥97.5% (HPLC)
C18H20N2O2
4, 4'-Bis(dimethylamino)benzil