missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Bis(diethylamino)benzophenone, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 170810250

 View more versions of this product

Catalog No. AC170810250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4, 4';-Bis(diethylamino)benzophenone
90-93-7
100.0
Glass bottle
≥99.0%
C21H28N2O
324.45
14, 98
93° to 95°C
99+%
99.0
Authentic
Crystalline Granules
99+%
[(C2H5)2NC6H4]2CO
MFCD00009044
Yellow to yellow-green
25g

Chemical Identifiers

90-93-7
100.0
C21H28N2O
324.45
4, 4';-Bis(diethylamino)benzophenone, 99+%
99.0
99+%
[(C2H5)2NC6H4]2CO
MFCD00009044
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 202-025-4

TSCA : TSCA