missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Bis(5-methyl-2-benzoxazolyl)stilbene, TCI America™

$111.53 - $334.59

Chemical Identifiers

CAS 2397-00-4
Molecular Formula C30H22N2O2
Molecular Weight (g/mol) 442.518
MDL Number MFCD00191316
InChI Key OKEZAUMKBWTTCR-AATRIKPKSA-N
PubChem CID 6436901
IUPAC Name 5-methyl-2-[4-[(E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=C(O5)C=CC(=C6)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B15645G View Documents TCI America
B15645G
5g Each for $111.53
 
B156425G View Documents TCI America
B156425G
25g Each for $334.59
 
Specifications

Chemical Identifiers

2397-00-4
442.518
OKEZAUMKBWTTCR-AATRIKPKSA-N
5-methyl-2-[4-[(E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
C30H22N2O2
MFCD00191316
6436901
CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=C(O5)C=CC(=C6)C

Specifications

Undesignated
2397-00-4
MFCD00191316
CC1=CC2=C(C=C1)OC(=N2)C3=CC=C(C=C3)C=CC4=CC=C(C=C4)C5=NC6=C(O5)C=CC(=C6)C
442.518
442.52
≥98.0% (N)
5g
C30H22N2O2
OKEZAUMKBWTTCR-AATRIKPKSA-N
5-methyl-2-[4-[(E)-2-[4-(5-methyl-1,3-benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-benzoxazole
6436901
Crystal-Powder at 20°C
4,4′-Bis(5-methyl-2-benzoxazolyl)stilbene
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0169

TSCA : Yes