Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Bis(2,5-dimethylstyryl)biphenyl, 99+%

$153.30 - $153.30

Chemical Identifiers

Chemical Name or Material 4,4'-Bis(2,5-dimethylstyryl)biphenyl
CAS 72814-85-8
Molecular Formula C32H30
Formula Weight 414.59
MDL Number MFCD00051223
Beilstein 6526037
Synonym BDB
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0918806

  SDS
alfa aesar™
L0918806
5g Each for $153.30
Description & Specifications

Specifications

Formula Weight 414.59
Assay ≥99%
Beilstein 6526037
Melting Point 196° to 200°C
MDL Number MFCD00051223
Chemical Name or Material 4,4'-Bis(2,5-dimethylstyryl)biphenyl
TSCA TSCA
Molecular Formula C32H30
CAS 72814-85-8
Synonym BDB
Quantity 5g