missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Bis(2,2-diphenylvinyl)biphenyl, TCI America™

$273.48 - $944.19

Chemical Identifiers

CAS 142289-08-5
Molecular Formula C40H30
Molecular Weight (g/mol) 510.68
MDL Number MFCD03093280
InChI Key UHXOHPVVEHBKKT-UHFFFAOYSA-N
Synonym DPVBi
PubChem CID 15475895
IUPAC Name 1-(2,2-diphenylethenyl)-4-[4-(2,2-diphenylethenyl)phenyl]benzene
SMILES C1=CC=C(C=C1)C(=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B27375G View Documents TCI America
B27375G
5g Each for $273.48
 
B273725G View Documents TCI America
B273725G
25g Each for $944.19
 
Specifications

Chemical Identifiers

142289-08-5
510.68
UHXOHPVVEHBKKT-UHFFFAOYSA-N
15475895
C1=CC=C(C=C1)C(=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
C40H30
MFCD03093280
DPVBi
1-(2,2-diphenylethenyl)-4-[4-(2,2-diphenylethenyl)phenyl]benzene

Specifications

Yellow
5g
C40H30
DPVBi
C1=CC=C(C=C1)C(=CC2=CC=C(C=C2)C3=CC=C(C=C3)C=C(C4=CC=CC=C4)C5=CC=CC=C5)C6=CC=CC=C6
510.68
510.68
≥98.0% (HPLC)
208°C
142289-08-5
MFCD03093280
UHXOHPVVEHBKKT-UHFFFAOYSA-N
1-(2,2-diphenylethenyl)-4-[4-(2,2-diphenylethenyl)phenyl]benzene
15475895
Crystal-Powder at 20°C
4,4′-Bis(2,2-diphenylvinyl)biphenyl
Safety and Handling

Safety and Handling

TSCA : No