missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Bipyridine, 98%

Manufacturer:  Alfa Aesar™ A1018706

 View more versions of this product

Catalog No. AAA1018706



Description

Description

Applications
4,4′-Bipyridine is used in transition-metal complex catalyst chemistry for uniform polymerization, in luminescence chemistry and in spectrophotometric analysis. It plays an important role as a photosensitizer and luminescent material. It is also used as a precursor to paraquat viz. N,N′-dimethyl-4,4′-bipyridinium.

Solubility
Soluble in ethanol, ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

4,4'-Bipyridine
98%
MFCD00006416
113176
111°C to 114°C
304°C to 305°C
MWVTWFVJZLCBMC-UHFFFAOYSA-N
4-pyridin-4-ylpyridine
11107
156.19
553-26-4
C10H8N2
UN2811
14,3348
4,4'-Bipyridyl; 4,4'-Dipyridyl
5g
C1=CN=CC=C1C2=CC=NC=C2
156.188
CHEBI:30985

Chemical Identifiers

553-26-4
156.188
MWVTWFVJZLCBMC-UHFFFAOYSA-N
11107
4-pyridin-4-ylpyridine
C10H8N2
MFCD00006416
4,4'-Bipyridyl; 4,4'-Dipyridyl
CHEBI:30985
C1=CN=CC=C1C2=CC=NC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement
P280-P301+P310-P361-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DW1760000

TSCA : TSCA

SDS Documents