Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4'-Bipiperidine dihydrochloride, 99%

$61.78 - $224.51

Chemical Identifiers

Chemical Name or Material 4,4'-Bipiperidine dihydrochloride
CAS 78619-84-8
Molecular Formula C10H20N2·2HCl
Formula Weight 241.21
MDL Number MFCD00005998
Beilstein 3687878
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1569606

  SDS
alfa aesar™
L1569606
5g Each for $61.78

AAL1569614

  SDS
alfa aesar™
L1569614
25g Each for $224.51
Description & Specifications

Specifications

Formula Weight 241.21
MDL Number MFCD00005998
Chemical Name or Material 4,4'-Bipiperidine dihydrochloride
Molecular Formula C10H20N2·2HCl
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
CAS 78619-84-8
Health Hazard 1 Warning
Quantity 5g
Beilstein 3687878
Melting Point >300°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.