missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Biphthalic Anhydride, TCI America™

$36.67 - $110.00

Chemical Identifiers

CAS 2420-87-3
Molecular Formula C16H6O6
Molecular Weight (g/mol) 294.218
MDL Number MFCD00039140
InChI Key WKDNYTOXBCRNPV-UHFFFAOYSA-N
Synonym Diphenyl-3,3′,4,4′-tetracarboxylic Dianhydride
PubChem CID 75494
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B132625G View Documents TCI America
B132625G
25g Each for $36.67
 
B1326100G View Documents TCI America
B1326100G
100g Each for $110.00
 
Specifications

Chemical Identifiers

2420-87-3
294.218
WKDNYTOXBCRNPV-UHFFFAOYSA-N
75494
C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
C16H6O6
MFCD00039140
Diphenyl-3,3′,4,4′-tetracarboxylic Dianhydride
5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione

Specifications

Yellow
25g
C16H6O6
Diphenyl-3,3′C,4,4′C-tetracarboxylic Dianhydride
C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
294.218
294.22
≥98.0% (T)
303°C
2420-87-3
MFCD00039140
WKDNYTOXBCRNPV-UHFFFAOYSA-N
5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione
75494
Crystal-Powder at 20°C
4,4′-Biphthalic Anhydride
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0833

TSCA : Yes