missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Biphenyldisulfonic Acid, TCI America™

$175.70 - $609.59

Chemical Identifiers

CAS 5314-37-4
Molecular Formula C12H10O6S2
Molecular Weight (g/mol) 314.326
MDL Number MFCD00035762
InChI Key ABSXMLODUTXQDJ-UHFFFAOYSA-N
PubChem CID 79202
IUPAC Name 4-(4-sulfophenyl)benzenesulfonic acid
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)O)S(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B16645G View Documents TCI America
B16645G
5g Each for $175.70
 
B166425G View Documents TCI America
B166425G
25g Each for $609.59
 
Specifications

Chemical Identifiers

5314-37-4
314.326
ABSXMLODUTXQDJ-UHFFFAOYSA-N
4-(4-sulfophenyl)benzenesulfonic acid
C12H10O6S2
MFCD00035762
79202
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)O)S(=O)(=O)O

Specifications

Undesignated
5314-37-4
MFCD00035762
ABSXMLODUTXQDJ-UHFFFAOYSA-N
4-(4-sulfophenyl)benzenesulfonic acid
79202
Crystal-Powder at 20°C
4,4′-Biphenyldisulfonic Acid
5g
C12H10O6S2
2585
C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)O)S(=O)(=O)O
314.326
314.33
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : Yes