missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Biphenyldicarboxaldehyde, TCI America™

$131.39 - $450.70

Chemical Identifiers

CAS 66-98-8
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
InChI Key FEHLIYXNTWAEBQ-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarboxaldehyde, 1,1'-biphenyl-4,4'-dicarbaldehyde, 4,4'-diformylbiphenyl, biphenyl-4,4'-dicarbaldehyde, 4,4'-biphenyldicarboxyaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, p,p'-diformylbiphenyl, 4-4-formylphenyl benzaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, 4,4'-bis formyl biphenyl
PubChem CID 6200
IUPAC Name 4-(4-formylphenyl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28541G View Documents TCI America
B28541G
1g Each for $131.39
 
B28545G View Documents TCI America
B28545G
5g Each for $450.70
 
Specifications

Chemical Identifiers

66-98-8
210.232
4,4'-biphenyldicarboxaldehyde, 1,1'-biphenyl-4,4'-dicarbaldehyde, 4,4'-diformylbiphenyl, biphenyl-4,4'-dicarbaldehyde, 4,4'-biphenyldicarboxyaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, p,p'-diformylbiphenyl, 4-4-formylphenyl benzaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, 4,4'-bis formyl biphenyl
4-(4-formylphenyl)benzaldehyde
C14H10O2
FEHLIYXNTWAEBQ-UHFFFAOYSA-N
6200
C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C=O

Specifications

White-Yellow
1g
C14H10O2
FEHLIYXNTWAEBQ-UHFFFAOYSA-N
4-(4-formylphenyl)benzaldehyde
6200
Crystal-Powder at 20°C
4,4′-Biphenyldicarboxaldehyde
146°C
66-98-8
4,4'-biphenyldicarboxaldehyde, 1,1'-biphenyl-4,4'-dicarbaldehyde, 4,4'-diformylbiphenyl, biphenyl-4,4'-dicarbaldehyde, 4,4'-biphenyldicarboxyaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, p,p'-diformylbiphenyl, 4-4-formylphenyl benzaldehyde, 1,1'-biphenyl-4,4'-dicarboxaldehyde, 4,4'-bis formyl biphenyl
C1=CC(=CC=C1C=O)C2=CC=C(C=C2)C=O
210.232
210.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : DV2800200

TSCA : No

Recommended Storage : Refrigerator