missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4′-Bicyclohexanone, TCI America™

$71.81 - $580.56

Chemical Identifiers

CAS 23391-99-3
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.274
MDL Number MFCD00210694
InChI Key OQIDYCVWAFZRTR-UHFFFAOYSA-N
Synonym 4,4′C-Dioxobicyclohexyl
PubChem CID 264825
IUPAC Name 4-(4-oxocyclohexyl)cyclohexan-1-one
SMILES C1CC(=O)CCC1C2CCC(=O)CC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B240525G View Documents TCI America
B240525G
25g Each for $71.81
 
B2405500G View Documents TCI America
B2405500G
500g Each for $580.56
 
Specifications

Chemical Identifiers

23391-99-3
194.274
OQIDYCVWAFZRTR-UHFFFAOYSA-N
264825
C1CC(=O)CCC1C2CCC(=O)CC2
C12H18O2
MFCD00210694
4,4′C-Dioxobicyclohexyl
4-(4-oxocyclohexyl)cyclohexan-1-one

Specifications

White-Yellow
25g
C12H18O2
4,4′C-Dioxobicyclohexyl
C1CC(=O)CCC1C2CCC(=O)CC2
194.274
194.27
≥98.0% (GC)
118°C
23391-99-3
MFCD00210694
OQIDYCVWAFZRTR-UHFFFAOYSA-N
4-(4-oxocyclohexyl)cyclohexan-1-one
264825
Crystal-Powder at 20°C
4,4′-Bicyclohexanone
Safety and Handling

Safety and Handling

TSCA : No