Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine, 95%

$77.59 - $175.47

Chemical Identifiers

Chemical Name or Material 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine
CAS 885952-23-8
Molecular Formula C15H16BrN3O2
Formula Weight 350.21
MDL Number MFCD06798240
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2691103

 
alfa aesar™
H2691103
1g Each for $175.47

AAH26911MD

 
alfa aesar™
H26911MD
250mg Each for $77.59
Description & Specifications

Specifications

Formula Weight 350.21
Chemical Name or Material 4-(4-Benzyloxy-5-bromo-2-pyrimidinyl)morpholine
Assay 95%
Molecular Formula C15H16BrN3O2
MDL Number MFCD06798240
CAS 885952-23-8
Melting Point 114° to 116°C
Quantity 1g