Please login to your online account to display your discounted pricing

4,4'-Azodianiline 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  401580050

 View more versions of this product

Catalog No. AC401580050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Powder
Formula Weight 212.25
Quantity 5g
Color Brown
EINECS Number 208-690-7
TSCA TSCA
RTECS Number BW7450000
Merck Index 15, 2979
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12H12N4
Packaging Glass bottle
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.
Chemical Name or Material 4, 4'-Azodianiline
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00041892
CAS 538-41-0
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
IF exposed or concerned: Get medical advice/attention.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4, 4'-Azodianiline
Name Note 95%
CAS 538-41-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12H12N4
Formula Weight 212.25
MDL Number MFCD00041892
Accessories

GHS Signal Word: Danger

GHS P Statement
IF exposed or concerned: Get medical advice/attention.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.

SDS & Safety Documents