missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 1571-75-1 |
---|---|
Molecular Formula | C20H18O2 |
Molecular Weight (g/mol) | 290.362 |
MDL Number | MFCD00134687 |
InChI Key | VOWWYDCFAISREI-UHFFFAOYSA-N |
Synonym | 1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4′C-(1-Phenylethylidene)diphenol |
PubChem CID | 623849 |
IUPAC Name | 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol |
SMILES | CC(C1=CC=CC=C1)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O |
Chemical Identifiers
1571-75-1 | |
290.362 | |
VOWWYDCFAISREI-UHFFFAOYSA-N | |
623849 | |
CC(C1=CC=CC=C1)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O |
C20H18O2 | |
MFCD00134687 | |
1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4′C-(1-Phenylethylidene)diphenol | |
4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol |
Specifications
White | |
25g | |
C20H18O2 | |
1,1-Bis(4-hydroxyphenyl)-1-phenylethane, 4,4′C-(1-Phenylethylidene)diphenol | |
CC(C1=CC=CC=C1)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O | |
290.362 | |
290.36 | |
≥98.0% (GC) |
189°C | |
1571-75-1 | |
MFCD00134687 | |
VOWWYDCFAISREI-UHFFFAOYSA-N | |
4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol | |
623849 | |
Crystal-Powder at 20°C | |
4,4′-(alpha-Methylbenzylidene)bisphenol |
Safety and Handling
EINECSNumber : (4)-1537
TSCA : No