Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Acetamidophenyl)-2-aminothiazole, 97%

$175.47 - $523.35

Chemical Identifiers

Chemical Name or Material 4-(4-Acetamidophenyl)-2-aminothiazole
CAS 21674-96-4
Molecular Formula C11H11N3OS
Formula Weight 233.29
MDL Number MFCD00207103
Synonym 4-(4-Acetamidophenyl)-2-thiazolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2785406

 
alfa aesar™
H2785406
5g Each for $175.47

AAH2785414

 
alfa aesar™
H2785414
25g Each for $523.35
Description & Specifications

Specifications

Formula Weight 233.29
Chemical Name or Material 4-(4-Acetamidophenyl)-2-aminothiazole
Assay 97%
Molecular Formula C11H11N3OS
MDL Number MFCD00207103
CAS 21674-96-4
Synonym 4-(4-Acetamidophenyl)-2-thiazolamine
Melting Point 248° to 252°C
Quantity 5g
Odor Odorless