missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane, 98%, stabilized, ACROS Organics™

Chemical Identifiers

CAS 4627-10-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H15BO2
Formula Weight 154.02
MDL Number MFCD00066915
Chemical Name or Material 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC164182500 SDS Thermo Fisher Scientific
AC164182500
Glass Bottle in Plastic Container 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC164180010 SDS Thermo Fisher Scientific
AC164180010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

4627-10-5
100.0
C8H15BO2
MFCD00066915
≥97.5%
97.5
98%
154.02
4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane

Specifications

154.02
250mg
Glass Bottle in Plastic Container
100.0
97.5
Authentic
MFCD00066915
4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
Liquid
Clear colorless
0.8800g/mL
98%
10% max. tetrahydrofuran
C8H15BO2
4627-10-5
≥97.5%