Please login to your online account to display your discounted pricing

4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane, 98%, stabilized, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC164180010

 View more versions of this product

Catalog No. AC164180010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.02
Physical Form Liquid
Assay ≥97.5%
Quantity 1g
Color Clear colorless
Density 0.8800g/mL
CAS Max % 100.0
Stabilizer 10% max. tetrahydrofuran
Molecular Formula C8H15BO2
Chemical Name or Material 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
MDL Number MFCD00066915
CAS 4627-10-5
CAS Min % 97.5
Health Hazard 2 GHS P Statement Flammable liquid and vapor. Suspected of causing cancer.
Packaging Glass bottle
Assay Percent Range 98%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood.

Specifications

Chemical Name or Material 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
CAS 4627-10-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H15BO2
Formula Weight 154.02
MDL Number MFCD00066915
Accessories

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Obtain special instructions before use. Do not handle until all safety precautions have been read and understood.

GHS P Statement Flammable liquid and vapor. Suspected of causing cancer.

SDS & Safety Documents