missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4,6-Dichloro-2-pyrimidinyl)morpholine, 97%

$155.14 - $464.16

Chemical Identifiers

CAS 10397-13-4
Molecular Formula C8H9Cl2N3O
Molecular Weight (g/mol) 234.08
MDL Number MFCD05022359
InChI Key OXCOCPRVQUEIOL-UHFFFAOYSA-N
Synonym 2-4-morpholino-4,6-dichloropyrimidine
PubChem CID 2772068
IUPAC Name 4-(4,6-dichloropyrimidin-2-yl)morpholine
SMILES C1COCCN1C2=NC(=CC(=N2)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54171MD View Documents Alfa Aesar™
H54171MD
250mg Each for $155.14
 
AAH5417103 View Documents Alfa Aesar™
H5417103
1g Each for $464.16
 
Specifications

Chemical Identifiers

10397-13-4
234.08
OXCOCPRVQUEIOL-UHFFFAOYSA-N
2772068
C1COCCN1C2=NC(=CC(=N2)Cl)Cl
C8H9Cl2N3O
MFCD05022359
2-4-morpholino-4,6-dichloropyrimidine
4-(4,6-dichloropyrimidin-2-yl)morpholine

Specifications

250mg
C8H9Cl2N3O
2-4-morpholino-4,6-dichloropyrimidine
C1COCCN1C2=NC(=CC(=N2)Cl)Cl
234.08
234.08
4-(4,6-Dichloro-2-pyrimidinyl)morpholine
10397-13-4
MFCD05022359
OXCOCPRVQUEIOL-UHFFFAOYSA-N
4-(4,6-dichloropyrimidin-2-yl)morpholine
2772068
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures