missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4,6-Dichloro-2-pyrimidinyl)morpholine, 97%

Manufacturer:  Alfa Aesar™ H5417103

 View more versions of this product

Catalog No. AAH5417103



Specifications

Specifications

4-(4,6-Dichloro-2-pyrimidinyl)morpholine
10397-13-4
MFCD05022359
OXCOCPRVQUEIOL-UHFFFAOYSA-N
4-(4,6-dichloropyrimidin-2-yl)morpholine
2772068
97%
1g
C8H9Cl2N3O
2-4-morpholino-4,6-dichloropyrimidine
C1COCCN1C2=NC(=CC(=N2)Cl)Cl
234.08
234.08

Chemical Identifiers

10397-13-4
234.08
OXCOCPRVQUEIOL-UHFFFAOYSA-N
2772068
C1COCCN1C2=NC(=CC(=N2)Cl)Cl
C8H9Cl2N3O
MFCD05022359
2-4-morpholino-4,6-dichloropyrimidine
4-(4,6-dichloropyrimidin-2-yl)morpholine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents