missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4',4''-Trimethyltriphenylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6416203

 View more versions of this product

Catalog No. AAH6416203



Description

Description

Used for proteomics research. Also employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for proteomics research. Also employed as a intermediate for pharmaceutical.

Solubility
Soluble in chloroform and tetrahydrofuran (THF).

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

4,4',4''-Trimethyltriphenylamine
114°C to 118°C (literature)
C21H21N
1 g
tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine
YXYUIABODWXVIK-UHFFFAOYSA-N
4-methyl-N,N-bis(4-methylphenyl)aniline
70873
98%
1159-53-1
White
MFCD00674043
2215309
Soluble in chloroform and tetrahydrofuran (THF).
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C
287.406
287.41
Solid

Chemical Identifiers

1159-53-1
287.406
YXYUIABODWXVIK-UHFFFAOYSA-N
70873
CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C
C21H21N
MFCD00674043
tri-p-tolylamine, 4,4',4-trimethyltriphenylamine, 4,4,4-trimethyltriphenylamine, tri p-tolyl amine, benzenamine, 4-methyl-n,n-bis 4-methylphenyl, 4-methyl-n,n-bis 4-methylphenyl aniline, tris 4-methylphenyl amine, n,n-di-p-tolyl-p-toluidine, n,n,n-tri p-tolyl amine, trip-tolylamine
4-methyl-N,N-bis(4-methylphenyl)aniline
Safety and Handling

Safety and Handling

EINECSNumber : 214-595-1

TSCA : Yes

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only