missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4',4''-Trimethoxytrityl chloride, 97%

$37.64 - $673.91

Chemical Identifiers

CAS 49757-42-8
Molecular Formula C22H21ClO3
Molecular Weight (g/mol) 368.857
MDL Number MFCD00008408
InChI Key OBFCMKPFILBCSQ-UHFFFAOYSA-N
Synonym 4,4',4-trimethoxytrityl chloride, 4,4',4-chloromethanetriyl tris methoxybenzene, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, 4,4',4'-trimethoxytrityl chloride, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, acmc-1an33, 4,4',4'-trimethoxytritylchloride, tris-p-methoxyphenyl chloromethane, 1-bis 4-methoxyphenyl chloromethyl-4-methoxybenzene, 1-chlorobis 4-methoxyphenyl methyl-4-methoxybenzene
PubChem CID 630505
IUPAC Name 1-[chloro-bis(4-methoxyphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0062903 View Documents Alfa Aesar™
L0062903
1g Each for $37.64
 
AAL0062906 View Documents Alfa Aesar™
L0062906
5g Each for $157.30
 
AAL0062914 View Documents Alfa Aesar™
L0062914
25g Each for $673.91
 
Specifications

Chemical Identifiers

49757-42-8
368.857
OBFCMKPFILBCSQ-UHFFFAOYSA-N
630505
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)Cl
C22H21ClO3
MFCD00008408
4,4',4-trimethoxytrityl chloride, 4,4',4-chloromethanetriyl tris methoxybenzene, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, 4,4',4'-trimethoxytrityl chloride, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, acmc-1an33, 4,4',4'-trimethoxytritylchloride, tris-p-methoxyphenyl chloromethane, 1-bis 4-methoxyphenyl chloromethyl-4-methoxybenzene, 1-chlorobis 4-methoxyphenyl methyl-4-methoxybenzene
1-[chloro-bis(4-methoxyphenyl)methyl]-4-methoxybenzene

Specifications

~150°C
49757-42-8
MFCD00008408
4,4',4-trimethoxytrityl chloride, 4,4',4-chloromethanetriyl tris methoxybenzene, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, 4,4',4'-trimethoxytrityl chloride, benzene, 1,1',1-chloromethylidyne tris 4-methoxy, acmc-1an33, 4,4',4'-trimethoxytritylchloride, tris-p-methoxyphenyl chloromethane, 1-bis 4-methoxyphenyl chloromethyl-4-methoxybenzene, 1-chlorobis 4-methoxyphenyl methyl-4-methoxybenzene
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)Cl
368.857
368.86
4,4',4''-Trimethoxytrityl chloride
1g
C22H21ClO3
892836
OBFCMKPFILBCSQ-UHFFFAOYSA-N
1-[chloro-bis(4-methoxyphenyl)methyl]-4-methoxybenzene
630505
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning