Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4',4''-Trimethoxytrityl alcohol, 95%

$49.11 - $485.18

Chemical Identifiers

Chemical Name or Material 4,4',4''-Trimethoxytrityl alcohol
CAS 3010-81-9
Molecular Formula C22H22O4
Formula Weight 350.41
MDL Number MFCD00014898
Beilstein 1890559
Synonym Tris(4-methoxyphenyl) carbinol; Tris(4-methoxyphenyl)methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2451603

  SDS
alfa aesar™
B2451603
1g Each for $49.11

AAB2451614

  SDS
alfa aesar™
B2451614
25g Each for $485.18
Description & Specifications

Specifications

Formula Weight 350.41
Chemical Name or Material 4,4',4''-Trimethoxytrityl alcohol
Assay 95%
Molecular Formula C22H22O4
MDL Number MFCD00014898
CAS 3010-81-9
Beilstein 1890559
Synonym Tris(4-methoxyphenyl) carbinol; Tris(4-methoxyphenyl)methanol
Melting Point 76° to 82°C
Quantity 1g