missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4',4''-Trihydroxytriphenylmethane, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC450101000

 View more versions of this product

Catalog No. AC450101000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4, 4', 4''-Trihydroxytriphenylmethane
244.0°C to 246.0°C
C19H16O3
Glass bottle
100g
WFCQTAXSWSWIHS-UHFFFAOYSA-N
4-[bis(4-hydroxyphenyl)methyl]phenol
69047
97%
603-44-1
96% min. (HPLC)
MFCD00191589
4,4',4-methanetriyltriphenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, 4,4',4-trihydroxytriphenylmethane, 4-bis 4-hydroxyphenyl methyl phenol, leucaurin, leucoaurin, leucoaurine, phenol, 4,4',4-methylidynetris, phenol, methylidynetris
Solubility in water: insoluble. Other solubilities: soluble in methanol
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
292.33
292.33

Chemical Identifiers

603-44-1
292.33
WFCQTAXSWSWIHS-UHFFFAOYSA-N
69047
OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
C19H16O3
MFCD00191589
4,4',4-methanetriyltriphenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, 4,4',4-trihydroxytriphenylmethane, 4-bis 4-hydroxyphenyl methyl phenol, leucaurin, leucoaurin, leucoaurine, phenol, 4,4',4-methylidynetris, phenol, methylidynetris
4-[bis(4-hydroxyphenyl)methyl]phenol
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-040-2

SDS
Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only