Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4,4-Trifluorocrotononitrile, 96%

Chemical Identifiers

Chemical Name or Material 4,4,4-Trifluorocrotononitrile
CAS 406-86-0
Molecular Formula CF3CH=CHCN
Formula Weight 121.06
MDL Number MFCD00190645
UN Number UN3276
Beilstein 6381034
Synonym 4,4,4-Trifluoro-2-butenenitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1687003

 
alfa aesar™
L1687003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 121.06
Chemical Name or Material 4,4,4-Trifluorocrotononitrile
CAS 406-86-0
Assay 96%
Molecular Formula CF3CH=CHCN
MDL Number MFCD00190645
Beilstein 6381034
Synonym 4,4,4-Trifluoro-2-butenenitrile
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S
UN Number UN3276
Health Hazard 2 GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 84° to 85°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H315-H319-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.