Learn More
4,4,4-Trifluorobutyric acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals B2218603
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
4,4,4-Trifluorobutyric acid | |
166°C to 167°C | |
MFCD00077604 | |
UN3261 | |
Soluble in water. | |
C(CC(F)(F)F)C(=O)O | |
142.077 | |
142.08 |
406-93-9 | |
C4H5F3O2 | |
1g | |
4,4,4-trifluorobutyric acid, 4,4,4-trifluoro-butyric acid, butanoic acid, 4,4,4-trifluoro, zlchem 266, acmc-1ai6e, 4,4,4-trifluorobutyricacid, trifluoropropylcarboxylic acid, 4,4,4 trifluorobutyric acid, ksc237i6h, 3-trifluoromethylpropionic acid | |
WTUCTMYLCMVYEX-UHFFFAOYSA-N | |
4,4,4-trifluorobutanoic acid | |
2777085 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only