missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4,4-Trifluorobutylamine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC432590010

 View more versions of this product

Catalog No. AC432590010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4, 4, 4-Trifluorobutylamine
1g
96.0
Authentic
Liquid
97%
127.11
Clear colorless
819-46-5
100.0
Glass bottle
≥96.0%
C4H8F3N

Chemical Identifiers

819-46-5
100.0
C4H8F3N
4, 4, 4-Trifluorobutylamine
96.0
97%
127.11
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger