Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4,4-Trifluoro-2-butanone, 97%

Chemical Identifiers

Chemical Name or Material 4,4,4-Trifluoro-2-butanone
CAS 2366-70-3
Molecular Formula CF3CH2COCH3
Formula Weight 126.08
MDL Number MFCD00077601
UN Number UN1224
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2151403

 
alfa aesar™
B2151403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2151406

 
alfa aesar™
B2151406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 126.08
Chemical Name or Material 4,4,4-Trifluoro-2-butanone
Assay 97%
Molecular Formula CF3CH2COCH3
MDL Number MFCD00077601
CAS 2366-70-3
UN Number UN1224
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Flash Point 37°C (99°F)
Boiling Point 95° to 96°C
Quantity 1g

Applications
It is used as a pharmaceutical intermediate. It is used for the synthesis of racemic 2-amino-5,5,5-trifluoro-3-methylpentanoic acid as fluorinated synthon.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges. Keep ignition sources away. Prevent formation of aerosols.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.