missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione, TCI America™

Manufacturer:  TCI America T158310G

 View more versions of this product

Catalog No. T158310G



Specifications

Specifications

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
74°C
893-33-4
MFCD00054501
WVVLURYIQCXPIV-UHFFFAOYSA-N
4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
70179
Crystal-Powder at 20°C
Undesignated
10g
C14H9F3O2
4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione, 1-2-naphthoyl-3,3,3-trifluoroacetone, 4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione, 3-2-naphthoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl, 2-naphthoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl, 4,4,4-trifluoro-3-oxo-2'-butyronaphthone, acmc-1bl5t
C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F
266.219
266.22
≥98.0% (GC)

Chemical Identifiers

893-33-4
266.219
WVVLURYIQCXPIV-UHFFFAOYSA-N
70179
C1=CC=C2C=C(C=CC2=C1)C(=O)CC(=O)C(F)(F)F
C14H9F3O2
MFCD00054501
4,4,4-trifluoro-1-naphthalen-2-yl butane-1,3-dione, 4,4,4-trifluoro-1-2-naphthyl-1,3-butanedione, 1-2-naphthoyl-3,3,3-trifluoroacetone, 4,4,4-trifluoro-1-2-naphthyl butane-1,3-dione, 3-2-naphthoyl-1,1,1-trifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthalenyl, 2-naphthoyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-naphthyl, 4,4,4-trifluoro-3-oxo-2'-butyronaphthone, acmc-1bl5t
4,4,4-trifluoro-1-naphthalen-2-ylbutane-1,3-dione
Safety and Handling

Safety and Handling

TSCA : Yes

Documents