Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,4',4''-Trichlorotrityl alcohol, 95%

$59.41 - $213.96

Chemical Identifiers

Chemical Name or Material 4,4',4''-Trichlorotrityl alcohol
CAS 3010-80-8
Molecular Formula C19H13Cl3O
Formula Weight 363.67
MDL Number MFCD00051795
Beilstein 2293156
Synonym Tris(p-chlorophenyl) carbinol; Tris(p-chlorophenyl)methanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0696504

 
alfa aesar™
L0696504
2g Each for $59.41

AAL0696509

 
alfa aesar™
L0696509
10g Each for $213.96
Description & Specifications

Specifications

Formula Weight 363.67
Chemical Name or Material 4,4',4''-Trichlorotrityl alcohol
CAS 3010-80-8
MDL Number MFCD00051795
Beilstein 2293156
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number PC4160000
Assay 95%
Molecular Formula C19H13Cl3O
Synonym Tris(p-chlorophenyl) carbinol; Tris(p-chlorophenyl)methanol
Health Hazard 1 Warning
Melting Point 94° to 97°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.