Learn More
4,4',4''-Trichlorotrityl alcohol, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0696509
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,4',4''-Trichlorotrityl alcohol | |
94°C to 97°C | |
MFCD00051795 | |
2293156 | |
BPFKTJMHOWDJKI-UHFFFAOYSA-N | |
tris(4-chlorophenyl)methanol | |
76379 | |
95% |
3010-80-8 | |
C19H13Cl3O | |
10 g | |
tris 4-chlorophenyl methanol, 4,4',4-trichlorotrityl alcohol, tris-4-chloro-phenyl-methanol, tri 4-chlorophenyl methanol, acmc-1clfy, tris-p-chlorophenyl methanol, tris 4-chlorophenyl methan-1-ol, 4,4,4-trichlorotrityl alcohol, benzenemethanol,4-chloro-a,a-bis 4-chlorophenyl | |
OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1 | |
363.66 | |
363.67 |
Chemical Identifiers
3010-80-8 | |
363.66 | |
BPFKTJMHOWDJKI-UHFFFAOYSA-N | |
76379 | |
OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1 |
C19H13Cl3O | |
MFCD00051795 | |
tris 4-chlorophenyl methanol, 4,4',4-trichlorotrityl alcohol, tris-4-chloro-phenyl-methanol, tri 4-chlorophenyl methanol, acmc-1clfy, tris-p-chlorophenyl methanol, tris 4-chlorophenyl methan-1-ol, 4,4,4-trichlorotrityl alcohol, benzenemethanol,4-chloro-a,a-bis 4-chlorophenyl | |
tris(4-chlorophenyl)methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 221-137-4
RTECSNumber : PC4160000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only