missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 269409-70-3
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.08
MDL Number MFCD02093756
InChI Key BICZJRAGTCRORZ-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxybenzeneboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacol cyclic ester, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacolate, 4-hydroxyphenylboronic acid, pinacol ester, 4-hydroxyphenylboronicacidpinacolester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 2734624
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366420050
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC366420050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

269409-70-3
220.08
BICZJRAGTCRORZ-UHFFFAOYSA-N
2734624
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
C12H17BO3
MFCD02093756
2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxybenzeneboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacol cyclic ester, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacolate, 4-hydroxyphenylboronic acid, pinacol ester, 4-hydroxyphenylboronicacidpinacolester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

269409-70-3
100.0
112°C to 117°C
Authentic
MFCD02093756
2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxybenzeneboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacol cyclic ester, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid pinacolate, 4-hydroxyphenylboronic acid, pinacol ester, 4-hydroxyphenylboronicacidpinacolester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
Solubility in water: insoluble in water
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
220.08
220.08
97%
96.0
Yellow to Brown
96% min. (GC)
C12H17BO3
Glass bottle
5g
BICZJRAGTCRORZ-UHFFFAOYSA-N
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
2734624
Crystalline Powder
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS
Product Certifications

RUO – Research Use Only