Please login to your online account to display your discounted pricing

4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide, ≥97%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC366370010

 View more versions of this product

Catalog No. AC366370010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Powder
Formula Weight 261.13
Quantity 1g
Color Light yellow to light beige
Melting Point 161° to 165°C
CAS Min % 97.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage Refrigerator +4°C
CAS 214360-60-8
MDL Number MFCD02093722
Assay Percent Range 97+%
Infrared Spectrum Authentic
Chemical Name or Material 4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide
Molecular Formula C14H20BNO3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide
CAS 214360-60-8
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C14H20BNO3
Formula Weight 261.13
MDL Number MFCD02093722
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Refrigerator +4°C

SDS & Safety Documents