missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 552846-17-0 |
---|---|
Molecular Formula | C14H23BN2O4 |
Molecular Weight (g/mol) | 294.16 |
MDL Number | MFCD05663873 |
InChI Key | IPISOFJLWYBCAV-UHFFFAOYSA-N |
Synonym | 1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid |
PubChem CID | 16217654 |
IUPAC Name | tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC384640010
|
Thermo Scientific Chemicals
384640010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC384640050
|
Thermo Scientific Chemicals
384640050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
552846-17-0 | |
294.16 | |
IPISOFJLWYBCAV-UHFFFAOYSA-N | |
16217654 | |
CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1 |
C14H23BN2O4 | |
MFCD05663873 | |
1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid | |
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate |
Specifications
552846-17-0 | |
100.0 | |
96% min. (HPLC) | |
C14H23BN2O4 | |
1g | |
Solubility in water: insoluble | |
CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1 | |
294.16 | |
294.16 | |
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester |
96.0 | |
Authentic | |
Glass bottle | |
MFCD05663873 | |
1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acid | |
IPISOFJLWYBCAV-UHFFFAOYSA-N | |
tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate | |
16217654 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only