Please login to your online account to display your discounted pricing

4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC74239CB

 View more versions of this product

Catalog No. CC74239CB

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 309.24
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine
Assay Percent Range 97%
Molecular Formula C15H24BNO3S
MDL Number MFCD11841079
CAS 364794-85-4
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-{[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)thien-2-yl]methyl}morpholine
CAS 364794-85-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H24BNO3S
Formula Weight 309.24
MDL Number MFCD11841079
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents