Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[4-(2-Chlorophenyl)-1H-pyrazol-1-yl]piperidine, 97%

$87.28 - $312.47

Chemical Identifiers

Chemical Name or Material 4-[4-(2-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
CAS 902836-42-4
Molecular Formula C14H16ClN3
Formula Weight 261.76
MDL Number MFCD08060999
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5080603

  SDS
alfa aesar™
H5080603
1g Each for $312.47

AAH50806MD

  SDS
alfa aesar™
H50806MD
250mg Each for $87.28
Description & Specifications

Specifications

Formula Weight 261.76
Chemical Name or Material 4-[4-(2-Chlorophenyl)-1H-pyrazol-1-yl]piperidine
Assay 97%
Molecular Formula C14H16ClN3
MDL Number MFCD08060999
CAS 902836-42-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.