missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4(3H)-Pyrimidone, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 228250100

 View more versions of this product

Catalog No. AC228250100



Description

Description

Specifications

Specifications

4(3H)-Pyrimidone
Glass bottle
163.0°C to 168.0°C
98+%
98.0
98+%
MFCD00167178
4 1h-pyrimidinone, 4 3h-pyrimidinone, 4 3h-pyrimidone, 4-hydroxypyrimidine, 4-pyrimidinol, 4-pyrimidinone, 4-pyrimidone, deaminoisocytosine, pyrimidin-4 3h-one, pyrimidin-4-ol
C1=CN=CNC1=O
96.09
96.09
98+%
Authentic
White to Yellow
10g
51953-17-4
100.0
C4H4N2O
24, 81
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
1H-pyrimidin-6-one
20695
Powder

Chemical Identifiers

51953-17-4
96.09
DNCYBUMDUBHIJZ-UHFFFAOYSA-N
20695
C1=CN=CNC1=O
C4H4N2O
MFCD00167178
4 1h-pyrimidinone, 4 3h-pyrimidinone, 4 3h-pyrimidone, 4-hydroxypyrimidine, 4-pyrimidinol, 4-pyrimidinone, 4-pyrimidone, deaminoisocytosine, pyrimidin-4 3h-one, pyrimidin-4-ol
1H-pyrimidin-6-one
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 257-545-4

TSCA : TSCA

SDS Documents