Please login to your online account to display your discounted pricing

4(3H)-Pyrimidone, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4(3H)-Pyrimidone
Name Note 98+%
CAS 51953-17-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00167178
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC228250100

  SDS
fisher scientific acros
228250100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 96.09
Quantity 10g
Color White to slightly yellow
TSCA TSCA
EINECS Number 257-545-4
Melting Point 163° to 168°C
Beilstein 24, 81
CAS Max % 100.0
CAS Min % 98.0
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Synonym 4-Hydroxypyrimidine
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C4H4N2O
MDL Number MFCD00167178
CAS 51953-17-4
Chemical Name or Material 4(3H)-Pyrimidone
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.