Please login to your online account to display your discounted pricing

Alfa Aesar™ 4(3H)-Pyrimidinone, 98%

$37.41 - $164.70

Chemical Identifiers

Chemical Name or Material 4 (3H)-Pyrimidinone
CAS 51953-17-4
Molecular Formula C4H4N2O
Formula Weight 96.09
MDL Number MFCD00167178
Beilstein 106975
Synonym 4-Hydroxypyrimidine; 4-Pyrimidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1085903

  SDS
alfa aesar™
A1085903
1g Each for $37.41

AAA1085906

  SDS
alfa aesar™
A1085906
5g Each for $164.70
Description & Specifications

Specifications

Formula Weight 96.09
Chemical Name or Material 4 (3H)-Pyrimidinone
TSCA TSCA
Assay 98%
Molecular Formula C4H4N2O
MDL Number MFCD00167178
CAS 51953-17-4
Beilstein 106975
Synonym 4-Hydroxypyrimidine; 4-Pyrimidinol
RTECS Number UW7350000
Melting Point 164° to 167°C
Quantity 1g