Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[3-(Trifluoromethyl)phenyl]-4-piperidinol, 97%

$69.12 - $203.19

Chemical Identifiers

Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-4-piperidinol
CAS 2249-28-7
Molecular Formula C12H14F3NO
Formula Weight 245.25
MDL Number MFCD00044832
Synonym 4-Hydroxy-4-[(3-trifluoromethyl)phenyl]piperidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2702506

 
alfa aesar™
H2702506
5g Each for $69.12

AAH2702514

 
alfa aesar™
H2702514
25g Each for $203.19
Description & Specifications

Specifications

Formula Weight 245.25
Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-4-piperidinol
Assay 97%
Molecular Formula C12H14F3NO
MDL Number MFCD00044832
CAS 2249-28-7
Synonym 4-Hydroxy-4-[(3-trifluoromethyl)phenyl]piperidine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 93° to 94°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.