Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
CAS 20069-30-1
Molecular Formula C8H8F3N3S
Formula Weight 235.23
MDL Number MFCD00041291
UN Number UN2811
Beilstein 991835
Synonym 4-(alpha,alpha,alpha-Trifluoro-m-tolyl)-3-thiosemicarbazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0929903

  SDS
alfa aesar™
L0929903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 235.23
CAS 20069-30-1
MDL Number MFCD00041291
Molecular Formula C8H8F3N3S
Assay 97%
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-[3-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Quantity 1g
Beilstein 991835
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
UN Number UN2811
Melting Point 120° to 122°C
Synonym 4-(alpha,alpha,alpha-Trifluoro-m-tolyl)-3-thiosemicarbazide
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.