missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide, TCI America™

Manufacturer:  TCI America T2819200MG

 View more versions of this product

Catalog No. T2819200MG



Specifications

Specifications

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Bromide
200mg
C9H6BrF3N2
3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
C1=CC(=CC=C1CBr)C2(N=N2)C(F)(F)F
279.06
279.06
≥95.0% (HPLC)
Undesignated
92367-11-8
3265
BCFUQXOZHPIAPS-UHFFFAOYSA-N
3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)diazirine
10826496
Clear Liquid at 20°C

Chemical Identifiers

92367-11-8
279.06
3-[4-(Bromomethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
3-[4-(bromomethyl)phenyl]-3-(trifluoromethyl)diazirine
C9H6BrF3N2
BCFUQXOZHPIAPS-UHFFFAOYSA-N
10826496
C1=CC(=CC=C1CBr)C2(N=N2)C(F)(F)F
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents