Please login to your online account to display your discounted pricing

4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00070CB

 View more versions of this product

Catalog No. MO00070CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 256.18
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
Assay Percent Range 97%
Molecular Formula C11H7F3N2O2
MDL Number MFCD03086203
CAS 220462-27-1
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]benzoic acid
CAS 220462-27-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H7F3N2O2
Formula Weight 256.18
MDL Number MFCD03086203
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents